Learn More
Ammonium Chloride/Hydroxide, 4 M, Ricca Chemical

$191.00 - $1406.00
Specifications
| Chemical Name or Material | Ammonium Chloride/Ammonium Hydroxide |
|---|---|
| Color | Colorless Liquid |
| CAS | 7732-18-5 , 12125-02-9 , 1336-21-6 |
| CAS Max % | 76.03, 20.57, 3.39 |
| Assay Percent Notes | Volumetric |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
R06274001A
![]() |
Ricca Chemical Company
R06274001A |
1 L |
Each for $191.00
|
|
|||||
R062740020F
![]() |
Ricca Chemical Company
R062740020F |
20 L |
Each for $1,406.00
|
|
|||||
Specifications
| Ammonium Chloride/Ammonium Hydroxide | |
| 7732-18-5 , 12125-02-9 , 1336-21-6 | |
| Volumetric | |
| Alkaline |
| Colorless Liquid | |
| 76.03, 20.57, 3.39 | |
| Laboratory | |
| Non-sterile |
Safety and Handling
ShelfLife : 12 months
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.