CAS RN 1336-21-6
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC Name: amine hydrate SMILES: N.O
Ammonia TS 2, Ricca Chemical
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC Name: amine hydrate SMILES: N.O
Ammonium Hydroxide ACS, MilliporeSigma™
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC Name: amine hydrate SMILES: N.O
Ammonium Hydroxide, 28.0 to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 SMILES: N.O
Ammonia R (175 g/L NH3), Ricca Chemical
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC Name: amine hydrate SMILES: N.O
Ammonium Hydroxide, 6.00 N, Ricca Chemical
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC Name: amine hydrate SMILES: N.O
Ammonium hydroxide, ACS reagent, 28-30% solution in water
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 SMILES: N.O
Ammonium hydroxide, 28% NH3
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC Name: azanium;hydroxide SMILES: N.O
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 SMILES: N.O
Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Ammonium hydroxide, extra pure, 25% solution in water
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC Name: amine hydrate SMILES: N.O
Buffer solution, pH 3.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Buffer solution pH 10.00 (+/-0.024 @ 25oC) Colored Blue Specpure NIST Traceable
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Ammonium hydroxide, for analysis, 25% solution in water
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 SMILES: N.O