Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium citrate dibasic

Catalog No. 251026X1KGR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-102-6X1KGR 6 x 1 kg
25-102-1KGR 1 kg
25-102-500GR 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25-102-6X1KGR Supplier Sigma Aldrich Supplier No. 251026X1KGR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ammonium citrate dibasic is generally used as a component of matrix solution in matrix-assisted laser desorption/ionization time-of-flight (maldi-tof) mass spectrometry analysis
  • It can also be used as both the carbon and nitrogen source to synthesize fluorescent nitrogen-doped carbon nanoparticles (cnps)

Specifications

CAS 3012-65-5
Molecular Formula C6H14N2O7
Linear Formula HOC(CO2H)(CH2CO2NH4)2
MDL Number MFCD00013068
Quantity 6 x 1 kg
Synonym Ammonium hydrogencitrate; Citric acid ammonium salt; Diammonium hydrogen citrate
Molecular Weight (g/mol) 226.18
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.