Learn More
Medchemexpress LLC Ammonium citrate dibasic | 3012-65-5 | MFCD00013068 | 99.0% | 100 G
SDP
Catalog No. 5000411167
Encompass_Preferred
Ammonium citrate dibasic, 99% (Ammonium hydrogencitrate, 99%) can be used as the carbon source to prepare carbon quantum dots (CDs). It is also a buffering agent.
  • Used as a carbon source to prepare carbon quantum dots (CDs)
  • Buffering agent
  • For research use only
Catalog No. 50-004-11167 Supplier Medchemexpress LLC Supplier No. HYB1529100G
Ammonium citrate dibasic, 99% (Ammonium hydrogencitrate, 99%) can be used as the carbon source to prepare carbon quantum dots (CDs). It is also a buffering agent.
  • Used as a carbon source to prepare carbon quantum dots (CDs)
  • Buffering agent
  • For research use only