Learn More
Medchemexpress LLC Ammonium citrate dibasic | 3012-65-5 | MFCD00013068 | 99.0% | 500 G
SDP
Catalog No. 5000412108
Encompass_Preferred
Ammonium citrate dibasic, 99% is a chemical compound suitable for various laboratory applications. It can be utilized as a carbon source for the preparation of carbon quantum dots (CDs) and also functions effectively as a buffering agent.
  • Can be used as the carbon source to prepare carbon quantum dots (CDs)
  • Functions as a buffering agent
Catalog No. 50-004-12108 Supplier Medchemexpress LLC Supplier No. HYB1529500G
Ammonium citrate dibasic, 99% is a chemical compound suitable for various laboratory applications. It can be utilized as a carbon source for the preparation of carbon quantum dots (CDs) and also functions effectively as a buffering agent.
  • Can be used as the carbon source to prepare carbon quantum dots (CDs)
  • Functions as a buffering agent