Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium cobalt(II) sulfate hexahydrate, 98%

Catalog No. AAA1062722
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1062722 100 g
AAA1062714 25 g
AAA1062736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1062722 Supplier Thermo Scientific Chemicals Supplier No. A1062722
Only null left
Add to Cart

CAS: 13586-38-4 | (NH4)2Co(SO4)2·6H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13586-38-4
Molecular Formula (NH4)2Co(SO4)2·6H2O
MDL Number MFCD00151049
Synonym Cobalt(II) ammonium sulfate; Cobaltous ammonium sulfate

Specifications

Melting Point ∼120°C (decomposition)
Quantity 100 g
UN Number UN3077
Formula Weight 395.21 (287.13 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 1.902
Chemical Name or Material Ammonium cobalt (II) sulfate hexahydrate
Hazard Category H302-H317-H350
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 237-043-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.