Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium cobalt(II) sulfate hexahydrate, 99%

Catalog No. p-7059074
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4390222 100 g
AA4390236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4390222 Supplier Thermo Scientific Chemicals Supplier No. 04390222
Only null left
Add to Cart

As spectroscopic standard, cobalt plating, catalysis, ceramics | CAS: 13586-38-4 | H8CoN2O8S2·6H2O

Ammonium cobalt(II) sulfate hexahydrate is uses as spectroscopic standard, cobalt plating, catalysis and ceramics. It is also used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13586-38-4
Molecular Formula H8CoN2O8S2·6H2O
MDL Number MFCD00151049
Synonym Cobalt(II) ammonium sulfate hexhydrate; Cobaltous ammonium sulfate hexahydrate

Specifications

Melting Point ∼120°C (decomposition)
Color Pink to Red
Physical Form Crystalline
Quantity 100 g
Linear Formula (NH4)2Co(SO4)2·6H2O
UN Number UN3077
Solubility Information Soluble in water, Insoluble in alcohol
Formula Weight 395.22 (287.14 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Sensitivity Hygroscopic
Density 1.902 g/mL
Chemical Name or Material Ammonium cobalt (II) sulfate hexahydrate
Show More Show Less
Hazard Category H302-H317-H350
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 237-043-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.