Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ammonium dihydrogen phosphate | 7722-76-1 | MFCD00003396 | 99.2% | 115.03 g/mol | NH4H2PO4 | 500 G
SDP

Catalog No. 5000301494
Encompass_Preferred

Ammonium dihydrogen phosphate is an inorganic salt used as a reagent, buffer component, and biochemical assay reagent in life-science research. It is supplied as a white crystalline powder and should be handled and stored according to the manufacturer's guidance to maintain stability and prevent moisture uptake.

  • Chemical formula: NH4H2PO4.
  • Molecular weight: 115.03 g/mol.
  • Purity: 99.2%.
  • Typical applications: reagent for biochemical assays and buffer preparation.
  • Physical form: white crystalline powder, soluble in water.
  • Storage: store sealed at 4°C and protect from moisture; solutions stable at -80°C (6 months) or -20°C (1 month).
  • Intended use: for research use only.

Catalog No. 50-003-01494 Supplier Medchemexpress LLC Supplier No. HYW019872500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ammonium dihydrogen phosphate is an inorganic salt used as a reagent, buffer component, and biochemical assay reagent in life-science research. It is supplied as a white crystalline powder and should be handled and stored according to the manufacturer's guidance to maintain stability and prevent moisture uptake.

  • Chemical formula: NH4H2PO4.
  • Molecular weight: 115.03 g/mol.
  • Purity: 99.2%.
  • Typical applications: reagent for biochemical assays and buffer preparation.
  • Physical form: white crystalline powder, soluble in water.
  • Storage: store sealed at 4°C and protect from moisture; solutions stable at -80°C (6 months) or -20°C (1 month).
  • Intended use: for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.