Learn More
Medchemexpress LLC Ammonium dihydrogen phosphate | 7722-76-1 | MFCD00003396 | 99.2% | 115.03 | NH4H2PO4 | 100 G
SDP
Catalog No. 5000301505
Encompass_Preferred
Ammonium dihydrogen phosphate (NH4H2PO4, CAS 7722-76-1) is an inorganic phosphate reagent supplied as a white crystalline powder for laboratory and analytical use. It serves as a buffer component and reagent in biochemical assays, chromatography, and other life-science applications.
  • High purity (99.2%) analytical-grade material.
  • Chemical formula NH4H2PO4 and molecular weight 115.03 g·mol-1.
  • White crystalline powder, soluble in water.
  • Suitable for buffer preparation, biochemical assays, and chromatography.
  • Available in common lab pack sizes including 100 g, 500 g, and 1 kg.
Catalog No. 50-003-01505 Supplier Medchemexpress LLC Supplier No. HYW019872100G
Ammonium dihydrogen phosphate (NH4H2PO4, CAS 7722-76-1) is an inorganic phosphate reagent supplied as a white crystalline powder for laboratory and analytical use. It serves as a buffer component and reagent in biochemical assays, chromatography, and other life-science applications.
  • High purity (99.2%) analytical-grade material.
  • Chemical formula NH4H2PO4 and molecular weight 115.03 g·mol-1.
  • White crystalline powder, soluble in water.
  • Suitable for buffer preparation, biochemical assays, and chromatography.
  • Available in common lab pack sizes including 100 g, 500 g, and 1 kg.