Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Dihydrogen Phosphate,, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1011265000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1011265000 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1011265000 Supplier MilliporeSigma Supplier No. 1.01126.5000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190°C
Color Colorless
pH 3.8 to 4.4
Physical Form Solid
Quantity 5 kg
Solubility Information 368g/L at 20°c
Formula Weight 115.02
Percent Purity >99.0 %
Grade ACS
Packaging Plastic Bottle
Vapor Pressure 0.066 hPa at 125°C
Density 1.80g/cm
Chemical Name or Material Ammonium Dihydrogen Phosphate
Show More Show Less
EINECSNumber 231-764-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.