Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium dihydrogenphosphate

Catalog No. 20400525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-400-525G 25 g
20-400-5500G 500 g
20-400-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-400-525G Supplier Sigma Aldrich Supplier No. 20400525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium dihydrogenphosphate

Specifications

Melting Point 190°C
Quantity 25 g
Molecular Formula H6NO4P
Linear Formula NH4H2PO4
MDL Number MFCD00003396
Synonym Ammonium phosphate monobasic; mono-Ammonium phosphate; prim-Ammonium phosphate; Ammonium dihydrogenphosphate
Molecular Weight (g/mol) 115.03
Percent Purity 99.999% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.