Learn More
Strem, An Ascensus Company CAS# 27546-07-2. 100g. Ammonium dimolybdate, 99%. MFCD00010882
SDP
Catalog No. 5090115319
Encompass
CAS# 27546-07-2. 100g. Ammonium dimolybdate, 99%. MFCD00010882. Molecular Weight: 339.95. Molecular Formula: (NH4)2Mo2O7. Color/form: white pwdr. Strem# 93-4201. http://www.strem.com/catalog/v/93-4201/
Catalog No. 50-901-15319 Supplier Strem, An Ascensus Company Supplier No. 934201100G
CAS# 27546-07-2. 100g. Ammonium dimolybdate, 99%. MFCD00010882. Molecular Weight: 339.95. Molecular Formula: (NH4)2Mo2O7. Color/form: white pwdr. Strem# 93-4201. http://www.strem.com/catalog/v/93-4201/