Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Fluoborate, MP Biomedicals

Catalog No. ICN204090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204090 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204090 Supplier MP Biomedicals, Inc Supplier No. 0520409005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium salt commonly used for research and further manufacturing applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-83-0
Molecular Formula BF4H4N
Molecular Weight (g/mol) 104.84
MDL Number MFCD06245785
InChI Key PDTKOBRZPAIMRD-UHFFFAOYSA-O
Synonym ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
PubChem CID 9964072
IUPAC Name ammonium tetrafluoroboranuide
SMILES [NH4+].F[B-](F)(F)F

Specifications

Physical Form Powder
Quantity 100 g
Linear Formula NH4BF4
Formula Weight 104.84
Chemical Name or Material Ammonium Fluoborate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.