Learn More
Strem, An Ascensus Company CAS# 12125-01-8. 500g. Ammonium fluoride, 98+% (ACS). MFCD00011423
SDP
Catalog No. 5090113568
Encompass
CAS# 12125-01-8. 500g. Ammonium fluoride, 98+% (ACS). MFCD00011423. Molecular Weight: 37.04. Molecular Formula: NH4F. Color/form: white xtl. Strem# 93-0212. http://www.strem.com/catalog/v/93-0212/
Catalog No. 50-901-13568 Supplier Strem, An Ascensus Company Supplier No. 930212500G
CAS# 12125-01-8. 500g. Ammonium fluoride, 98+% (ACS). MFCD00011423. Molecular Weight: 37.04. Molecular Formula: NH4F. Color/form: white xtl. Strem# 93-0212. http://www.strem.com/catalog/v/93-0212/