Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Fluoride, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCA0440500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCA0440500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCA0440500 Supplier Ricca Chemical Company Supplier No. RDCA0440500A6
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

CAS Max % 100
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity min 98%
Grade ACS
Packaging Natural Poly Bottle
Residue after Ignition max. 0.01%
Chemical Name or Material Ammonium Fluoride
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.