Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium fluoride solution, Solstice

Catalog No. 60090121 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-121 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-121 Supplier Solstice Supplier No. 40292H5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Semiconductor grade VLSI ( 17600), 40%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 12125-01-8, 7732-18-5
Boiling Point > 100°C (1.013 hPa)
CAS Min % ≥30.00%, ≥50.00%
CAS Max % <50.00%, <70.00%
Purity Grade Notes Semiconductor grade reagent
Molecular Weight (g/mol) 37.04
Packaging Plastic Can
Quantity 5 L
Physical Form Liquid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.