Learn More
Selleck Chemical LLC Ammonium formate
SDP
Catalog No. 5000003380
Encompass_Preferred
Ammonium Formate is the ammonium salt of formic acid It is widely used in various organic reactions
Catalog No. 50-000-03380 Supplier Selleck Chemical LLC Supplier No. S626225MG
Ammonium Formate is the ammonium salt of formic acid It is widely used in various organic reactions