Learn More
Waters Corp Waters Corporation Ammonium Formate Solution, Glycan Analysis, 50 mM, Mobile Phase Concentrate
Catalog No. 50596969 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This concentrated 50 mM ammonium formate buffer is designed for use with the Biopharm/Glycan system, UPLC H-class, and ACQUITY UPLC BEH Glycan Column. It is optimized for released N-glycan analysis, offering increased speed of deglycosylation and streamlined labeling within a smart workflow for rapid sample preparation and enhanced FLR and MS response.
  • Concentrated form (10 mL of 5050 mM ammonium formate in water with 3.8% formic acid matrix).
  • Direct dilution capability to 50 mM at pH 4.4.
  • Optimized for released N-glycan analysis.
  • Compatible with Glycoworks system.
  • Allows sample cleanup in as little as 30 minutes.
Catalog No. 50-596-969 Supplier Waters Corp Supplier No. 186007081
This concentrated 50 mM ammonium formate buffer is designed for use with the Biopharm/Glycan system, UPLC H-class, and ACQUITY UPLC BEH Glycan Column. It is optimized for released N-glycan analysis, offering increased speed of deglycosylation and streamlined labeling within a smart workflow for rapid sample preparation and enhanced FLR and MS response.
  • Concentrated form (10 mL of 5050 mM ammonium formate in water with 3.8% formic acid matrix).
  • Direct dilution capability to 50 mM at pH 4.4.
  • Optimized for released N-glycan analysis.
  • Compatible with Glycoworks system.
  • Allows sample cleanup in as little as 30 minutes.