Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium formate solution, 540-69-2, MFCD00013103, 100 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 78314100MLF
Encompass
Linear Formula: HCOONH4, Molecular Weight: 63.06, BioUltra, 10 M in H2O.
Catalog No. NC1134139