Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium formate Solution, For HPLC, 10 mM in Water, MilliporeSigma™ Supelco™

Catalog No. 111014542 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4542 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4542 Supplier MilliporeSigma Supelco Supplier No. 7146904X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Food and Beverages
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00013103

Specifications

Melting Point -119°C to +121°C
CAS 540-69-2
Density 0.998 g/mL (at 25°C)
Packaging Glass Bottle
Linear Formula HCOONH4
Quantity 4 x 4 L
Formula Weight 63.06
Grade For HPLC
Concentration or Composition (by Analyte or Components) 10 mM aq. soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.