Learn More
Selleck Chemical LLC Ammonium Glycyrrhizinate 5mg 1407-03-0 AMGZ, Glycyram, Glycyrrhizin ammonium salt, Glycyrrhizic acid ammonium salt
SDP
Catalog No. 505308
Encompass_Preferred
Ammonium Glycyrrhizinate (AMGZ, Glycyram, Glycyrrhizin ammonium salt, Glycyrrhizic acid ammonium salt) is a unique product from Licorice root with a very high sweetness. Ammonium glycyrrhizinate is a direct inhibitor of HMGB1 with anti-inflammatory, anti-tumor, anti-diabetic activities. Ammonium Glycyrrhizinate inhibits growth and cytopathology of several unrelated DNA and RNA viruses. Ammonium Glycyrrhizinate also inhibits both hepatic delta 4-5-reductase and 11 beta-hydroxysteroid dehydrogenase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-530-8 Supplier Selleck Chemical LLC Supplier No. S23765MG
Ammonium Glycyrrhizinate (AMGZ, Glycyram, Glycyrrhizin ammonium salt, Glycyrrhizic acid ammonium salt) is a unique product from Licorice root with a very high sweetness. Ammonium glycyrrhizinate is a direct inhibitor of HMGB1 with anti-inflammatory, anti-tumor, anti-diabetic activities. Ammonium Glycyrrhizinate inhibits growth and cytopathology of several unrelated DNA and RNA viruses. Ammonium Glycyrrhizinate also inhibits both hepatic delta 4-5-reductase and 11 beta-hydroxysteroid dehydrogenase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.