Learn More
Strem, An Ascensus Company CAS# 16919-58-7. 1g. Ammonium hexachloroplatinate(IV), 99%. MFCD00010886
SDP
Catalog No. 5090116355
Encompass
CAS# 16919-58-7. 1g. Ammonium hexachloroplatinate(IV), 99%. MFCD00010886. Molecular Weight: 443.89. Molecular Formula: (NH4)2PtCl6. Color/form: yellow pwdr. Strem# 93-7801. http://www.strem.com/catalog/v/93-7801/
Catalog No. 50-901-16355 Supplier Strem, An Ascensus Company Supplier No. 9378011G
CAS# 16919-58-7. 1g. Ammonium hexachloroplatinate(IV), 99%. MFCD00010886. Molecular Weight: 443.89. Molecular Formula: (NH4)2PtCl6. Color/form: yellow pwdr. Strem# 93-7801. http://www.strem.com/catalog/v/93-7801/