Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hexafluoroaluminate, 98%

Catalog No. AAB2058218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2058218 50 g
AAB2058209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2058218 Supplier Thermo Scientific Chemicals Supplier No. B2058218
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7784-19-2 | AlF6H12N3 | 195.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-19-2
Molecular Formula AlF6H12N3
Molecular Weight (g/mol) 195.09
MDL Number MFCD00042144
InChI Key OYHBNKHFKHBTRQ-UHFFFAOYSA-K
Synonym ammonium hexafluoroaluminate, ammonium cryolite, ammonium fluoaluminate, ammonium aluminum fluoride, triammonium hexafluoroaluminate, triammonium aluminum hexafluoride, triammoniumhexafluoroaluminate 3-, hexafluoroaluminate 3-triammonium, aluminum ammonium fluoride al nh4 3f6, ammonium hexafluoroaluminate nh4 3alf6
PubChem CID 24565
IUPAC Name triazanium;hexafluoroaluminum(3-)
SMILES [NH4+].[NH4+].[NH4+].F[Al-3](F)(F)(F)(F)F

Specifications

Quantity 50 g
UN Number UN3288
Merck Index 14,524
Formula Weight 195.09
Percent Purity 98%
Odor Odorless
Chemical Name or Material Ammonium hexafluoroaluminate
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 232-052-7
RTECSNumber BD0050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.