Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium hexafluorophosphate

Catalog No. 2165931KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-659-31KG 1 kg
21-659-325G 25 g
21-659-3100G 100 g
21-659-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-659-31KG Supplier Sigma Aldrich Supplier No. 2165931KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium hexafluorophosphate

Specifications

Melting Point 58°C
CAS 16941-11-0
Quantity 1 kg
Molecular Formula H4F6NP
Linear Formula NH4PF6
MDL Number MFCD00064642
Synonym Azanium hexafluorophosphate
Molecular Weight (g/mol) 163
Percent Purity 99.98% trace metals basis
Density 2.18 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.