Learn More
Strem, An Ascensus Company CAS# 16919-19-0. 500g. Ammonium hexafluorosilicate, min. 98%. MFCD00010887
SDP
Catalog No. 5090114167
Encompass
CAS# 16919-19-0. 500g. Ammonium hexafluorosilicate, min. 98%. MFCD00010887. Molecular Weight: 178.14. Molecular Formula: (NH4)2SiF6. Color/form: white pwdr. Strem# 93-1476. http://www.strem.com/catalog/v/93-1476/
Catalog No. 50-901-14167 Supplier Strem, An Ascensus Company Supplier No. 931476500G
CAS# 16919-19-0. 500g. Ammonium hexafluorosilicate, min. 98%. MFCD00010887. Molecular Weight: 178.14. Molecular Formula: (NH4)2SiF6. Color/form: white pwdr. Strem# 93-1476. http://www.strem.com/catalog/v/93-1476/