Learn More
Strem, An Ascensus Company CAS# 7783-28-0. 250g. Ammonium hydrogen phosphate, 98+% (ACS). MFCD00010891
SDP
Catalog No. 5090113571
Encompass
CAS# 7783-28-0. 250g. Ammonium hydrogen phosphate, 98+% (ACS). MFCD00010891. Molecular Weight: 132.06. Molecular Formula: (NH4)2HPO4. Color/form: white pwdr. Strem# 93-0220. http://www.strem.com/catalog/v/93-0220/
Catalog No. 50-901-13571 Supplier Strem, An Ascensus Company Supplier No. 930220250G
CAS# 7783-28-0. 250g. Ammonium hydrogen phosphate, 98+% (ACS). MFCD00010891. Molecular Weight: 132.06. Molecular Formula: (NH4)2HPO4. Color/form: white pwdr. Strem# 93-0220. http://www.strem.com/catalog/v/93-0220/