Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 1.00 Normal Standardized, NIST Traceable, Reagents

Catalog No. 01336154
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-154 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-154 Supplier Reagents Holdings Llc Supplier No. CA0025001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Hydroxide, 1.00 Normal Standardized, NIST Traceable, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonium Hydroxide, 1.00 Normal Standardized, NIST Traceable
pH 11.6
Color Colorless
Boiling Point 100°C
CAS 7732-18-5, 1336-21-6
Molecular Formula NH4OH/H2O
Synonym Aqua Ammonia
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.9925
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.