Learn More
Hach Company Ammonium Hydroxide, 500 mL
Supplier:  Hach Company 10649
Encompass
Ammonium Hydroxide ACS Grade, CAS No. 1336-21-6 NH4OH, approx. 28%, FW = 35.05. 500 mL Bottle
Catalog No. 50-199-4164