Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Ammonium Hydroxide
SDPSCI_ED

Catalog No. S72385 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72385 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72385 Supplier LaMotte™ Supplier No. 6537H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Hydroxide is a replacement reagent.

  • Ammonium Hydroxide comes as a kit size bottle.

Specifications

Color Clear Colorless
Composition 1336-21-6, 7732-18-5
Description Ammonium hydroxide
Volume (Metric) 60 mL
Format Liquid
Quantity 60 mL
Type Reagent
Volume (English) 2 oz.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.