Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide ACS AR, Macron Fine Chemicals™

Catalog No. MK1177212 Shop All Avantor Products
Change view
Click to view available options
SAFEMOR; 2.5L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-1177-212 SAFEMOR; 2.5L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-1177-212 Supplier Avantor Supplier No. 117746

Specifications

CAS 1336-21-6
Molecular Formula H5NO
MDL Number MFCD00066650
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
Quantity 2.5 L
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.05
Grade ACS
Packaging SAFEMOR
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.