Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide-Ammonium Chloride Buffer (R-20), Reagents

Catalog No. 01336197
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-197 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-197 Supplier Reagents Holdings Llc Supplier No. C83015NE500C
Only null left
Add to Cart
Add to Cart

Ammonium Hydroxide-Ammonium Chloride Buffer (R-20), Reagent, use for EDTA titrations. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonium Hydroxide-Ammonium Chloride Buffer (R-20)
CAS 7732-18-5, 1336-21-6, 1336-21-6
Color Colorless
For Use With (Application) Titration
pH >11.5
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Glass Bottle
Specific Gravity 1.0001
Sterility Non-sterile
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.