Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Ammonium Hydroxide Reagent
SDP

Catalog No. 22021398
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-398 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-398 Supplier StatLab Supplier No. SPA1343
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Ammonium Hydroxide Reagent - used as a bluing reagent for hematoxylin staining

  • The reagent is used to change soluble hematoxylin into an insoluble blue form, enhancing the contrast and clarity of nuclear staining
  • It is typically used after the hematoxylin staining and clearing/differentiation steps to achieve the desired staining intensity
  • It is a strong oxidizer and corrosive, and should be handled with caution
  • A MasterTech Special Stain Kit Component

Specifications

Grade Chemical Solution
Product Type Bluing Reagent
Quantity 500 mL
Solution Type Stain Kit
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.