Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium hydroxide solutio1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2212281LPCA
Encompass
Ammonium hydroxide is a weak base mainly used as a source to prepare other ammonium compounds like urea ammonium carbonate and ammonium sulfate etc.
Catalog No. 50-289-8881