Learn More
Bioworld Ammonium Hydroxide, 20% (v/v), 500 mL
Catalog No. 50255504
Encompass_Preferred
Synonym: Ammonia aqueous, Ammonia waterA complexant and base for traditional qualitative inorganic analysisHaz Mat Fee AppliesAppearance: LiquidBoiling Point : 38-100 °C at 760 mmHgDensity : 0.9 g/mL at 25 °C (lit.)HS Code : 28142000Melting Point: -60 °CRTECS Number: BQ9625000
Catalog No. 50-255-504 Supplier Bioworld Supplier No. 402201061
Synonym: Ammonia aqueous, Ammonia waterA complexant and base for traditional qualitative inorganic analysisHaz Mat Fee AppliesAppearance: LiquidBoiling Point : 38-100 °C at 760 mmHgDensity : 0.9 g/mL at 25 °C (lit.)HS Code : 28142000Melting Point: -60 °CRTECS Number: BQ9625000