Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Iodide, puriss., Solstice

Catalog No. 6001807 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-018-07 1 kg
60-044-508 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-018-07 Supplier Solstice Supplier No. 031011KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
SMILES [NH4+].[I-]

Specifications

Quantity 1 kg
Linear Formula NH4I
UN Number NONH for all modes of transport
IUPAC Name azanium;iodide
Formula Weight 144.94g/mol
Percent Purity ≥99%
Grade puriss.
Residue after Ignition ≤0.1% (as SO4)
Vapor Pressure 1mmHg (210.9°C)
Chemical Name or Material Ammonium Iodide
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H315-H319-H335
EINECSNumber 234-717-7
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.