Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium ion Solution for ISE 1,000 mg/kg N, MilliporeSigma™ Supelco™

Catalog No. 111012761 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2761 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2761 Supplier MilliporeSigma Supelco Supplier No. 09685250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 12125-02-9
Density 1.00 g/mL (at 20°C)
Concentration 1000 mg/kg N
Linear Formula NH4 Cl
MDL Number MFCD00143604
For Use With (Equipment) Ion Selective Electrode
Synonym Ammonium chloride solution
Type Analytical Standard
Quantity 250 mL
Formula Weight 53.49
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.