Learn More
Hach Company Ammonium Ionic Strength Adjustor (ISA) Powder Pillows, pk/100
Catalog No. 501996997 Shop All Hach Company Products
Encompass
Ionic Strength Adjustor (ISA) for Ammonium determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Ammonium level readings. Eliminate most interferences. Pack of 100 powder pillows.
Catalog No. 50-199-6997 Supplier Hach Company Supplier No. 2980699
Ionic Strength Adjustor (ISA) for Ammonium determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Ammonium level readings. Eliminate most interferences. Pack of 100 powder pillows.