Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium iron(II) sulfate hexahydrate

Catalog No. 2035055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-350-55G 5 g
20-350-5100G 100 g
20-350-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-350-55G Supplier Sigma Aldrich Supplier No. 2035055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium iron(II) sulfate hexahydrate

Specifications

Melting Point 100°C
CAS 7783-85-9
Quantity 5 g
Molecular Formula H8FeN2O8S2 -+ 6H2O
Linear Formula (NH4)2 Fe(SO4)2 -+ 6H2O
MDL Number MFCD00150530
Synonym Ammonium ferrous sulfate hexahydrate; Ammonium iron(II) sulfate; Mohrs salt
Molecular Weight (g/mol) 392.14
Percent Purity 99.997% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.