Learn More
Apexbio Technology LLC Ammonium iron(III) citrate 1185-57-5 1g
SDP
Catalog No. 5000094901
Encompass_Preferred
Ammonium iron(III) citrate (CAS 1185-57-5) is a coordination complex that serves as a bioavailable source of ferric ions It participates in cellular iron delivery supporting various enzymatic processes and redox reactions In experimental contexts it is often employed to modulate iron concentrations in biological systems and to induce oxidative stress Notably co-administration with metal chelators such as dimercaprol can increase the risk of renal toxicity reflecting potential interactions in metal homeostasis studies Ammonium iron(III) citrate is widely used in studies investigating iron metabolism oxidative damage and chelation therapies
Catalog No. 50-000-94901 Supplier Apexbio Technology LLC Supplier No. A47821000
Ammonium iron(III) citrate (CAS 1185-57-5) is a coordination complex that serves as a bioavailable source of ferric ions It participates in cellular iron delivery supporting various enzymatic processes and redox reactions In experimental contexts it is often employed to modulate iron concentrations in biological systems and to induce oxidative stress Notably co-administration with metal chelators such as dimercaprol can increase the risk of renal toxicity reflecting potential interactions in metal homeostasis studies Ammonium iron(III) citrate is widely used in studies investigating iron metabolism oxidative damage and chelation therapies