Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ammonium iron(III) sulfate dodecahydrate | 7783-83-7 | MFCD00149454 | 99.4% | 482.19 g·mol⁻¹ | NH4Fe(SO4)2·12H2O | 100 G
SDP

Catalog No. 5000299585
Encompass_Preferred

Ammonium iron(III) sulfate dodecahydrate (CAS 7783-83-7) is an inorganic salt supplied at high purity for laboratory and analytical applications. It is commonly used as a source of Fe(III) for preparing reagent and standard solutions in quantitative analysis.

  • High purity: 99.4%.
  • Molecular formula: NH4Fe(SO4)2·12H2O.
  • Molecular weight: 482.19 g·mol⁻¹.
  • Solid, crystalline form for easy handling and storage.
  • Used to prepare reagent and standard iron(III) solutions.
  • SDS and certificate of analysis available.
  • Packaging sizes include 100 g and 500 g; bulk quantities available.

Catalog No. 50-002-99585 Supplier Medchemexpress LLC Supplier No. HYW019927100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ammonium iron(III) sulfate dodecahydrate (CAS 7783-83-7) is an inorganic salt supplied at high purity for laboratory and analytical applications. It is commonly used as a source of Fe(III) for preparing reagent and standard solutions in quantitative analysis.

  • High purity: 99.4%.
  • Molecular formula: NH4Fe(SO4)2·12H2O.
  • Molecular weight: 482.19 g·mol⁻¹.
  • Solid, crystalline form for easy handling and storage.
  • Used to prepare reagent and standard iron(III) solutions.
  • SDS and certificate of analysis available.
  • Packaging sizes include 100 g and 500 g; bulk quantities available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.