Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iron(III) sulfate dodecahydrate, MilliporeSigma™

Catalog No. M1037760500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Packaging:
Fibre carton
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1037760500 500 g Plastic Bottle
M1037761000 1 kg Plastic Bottle
M1037765000 5 kg Plastic Bottle
M1037769050 50 kg Fibre carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1037760500 Supplier MilliporeSigma Supplier No. 1.03776.0500
Add to Cart
Add to Cart

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Quantity 500 g
Solubility Information Soluble in water: 1240g/L at 25°C
Formula Weight 482.19
Odor Odorless
Packaging Plastic Bottle
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate
EINECSNumber 233-382-4
Recommended Storage Store below 15°C (59°F), Tightly closed. Dry. Protected from Light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.