Learn More
Apexbio Technology LLC AMMONIUM-LACTATE-50G
SDP
Catalog No. 5000568277
Encompass_Preferred
Ammonium lactate (CAS No 515-98-0) is commonly studied in dermatological research for its moisturizing and keratolytic properties Originating from the neutralization of lactic acid with ammonium hydroxide it acts by increasing skin hydration and facilitating the exfoliation of dead skin cells In vitro its activity is observed in the millimolar range It is frequently used in cell models and animal studies to investigate skin barrier repair and potential as a topical agent Its effectiveness can be influenced by concentration which is tailored based on research design specifics
Catalog No. 50-005-68277 Supplier Apexbio Technology LLC Supplier No. C639550000
Ammonium lactate (CAS No 515-98-0) is commonly studied in dermatological research for its moisturizing and keratolytic properties Originating from the neutralization of lactic acid with ammonium hydroxide it acts by increasing skin hydration and facilitating the exfoliation of dead skin cells In vitro its activity is observed in the millimolar range It is frequently used in cell models and animal studies to investigate skin barrier repair and potential as a topical agent Its effectiveness can be influenced by concentration which is tailored based on research design specifics