Learn More
Strem, An Ascensus Company CAS# 12028-48-7. 100g. Ammonium metatungstate hydrate (99.9+%-W). MFCD00150662
SDP
Catalog No. 5090116300
Encompass
CAS# 12028-48-7. 100g. Ammonium metatungstate hydrate (99.9+%-W). MFCD00150662. Molecular Weight: 2956.45. Molecular Formula: (NH4)6H2W12O40∙XH2O. Color/form: white pwdr. Strem# 93-7420. http://www.strem.com/catalog/v/93-7420/
Catalog No. 50-901-16300 Supplier Strem, An Ascensus Company Supplier No. 937420100G
CAS# 12028-48-7. 100g. Ammonium metatungstate hydrate (99.9+%-W). MFCD00150662. Molecular Weight: 2956.45. Molecular Formula: (NH4)6H2W12O40∙XH2O. Color/form: white pwdr. Strem# 93-7420. http://www.strem.com/catalog/v/93-7420/