Learn More
Strem, An Ascensus Company CAS# 7803-55-6. 5g. Ammonium metavanadate (99.995%-V) PURATREM. MFCD00011430
SDP
Catalog No. 5090108582
Encompass
CAS# 7803-55-6. 5g. Ammonium metavanadate (99.995%-V) PURATREM. MFCD00011430. Molecular Weight: 116.98. Molecular Formula: NH4VO3. Color/form: white pwdr. Strem# 02-0700. http://www.strem.com/catalog/v/02-0700/
Catalog No. 50-901-08582 Supplier Strem, An Ascensus Company Supplier No. 0207005G
Add to Cart
Add to Cart
CAS# 7803-55-6. 5g. Ammonium metavanadate (99.995%-V) PURATREM. MFCD00011430. Molecular Weight: 116.98. Molecular Formula: NH4VO3. Color/form: white pwdr. Strem# 02-0700. http://www.strem.com/catalog/v/02-0700/