Learn More
Electron Microscopy Sciences Ammonium Molybdate 100 GR
SDP
Catalog No. 50980177
Encompass_Preferred
CAS #12027-67-7 (NH₄)₆MO₇O₂₄ ·4H₂O F.W. 1235.86
Used in the negative staining of viruses for TEM. Modifications caused by uncoupling agents (Muscatello, U., et al, J. Ultrastruct., Res., 52, 2 1975).
Catalog No. 50-980-177 Supplier Electron Microscopy Sciences Supplier No. 10500
CAS #12027-67-7 (NH₄)₆MO₇O₂₄ ·4H₂O F.W. 1235.86
Used in the negative staining of viruses for TEM. Modifications caused by uncoupling agents (Muscatello, U., et al, J. Ultrastruct., Res., 52, 2 1975).