Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium molybdate

Catalog No. 2779085G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
20 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-790-85G 5 g
27-790-8100G 100 g
27-790-820G 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 27-790-85G Supplier Sigma Aldrich Supplier No. 2779085G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium molybdate can be prepared by dissolving molybdenum trioxide in ammonia.Ammonium molybdate has been used to prepare phosphomolybdates. (NH4)2MoO4 was used as an analytical reagent for quantitative analysis of phosphates, silicates, arsenates and lead in their aqueous solutions.2 At saturated concentration, (NH4)2MoO4 was used to perform cryo-negative staining and as a negative stain in biological electron microscopy. Ammonium molybdate was used in study of crystallisation of cell-free-expressed membrane proteins.

Specifications

CAS 13106-76-8
Quantity 5 g
Molecular Formula H8MoN2O4
Linear Formula (NH4)2 MoO4
MDL Number MFCD00011596
Synonym Ammonium orthomolybdate; Diammonium molybdate
Molecular Weight (g/mol) 196.01
Percent Purity 99.98% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.