Learn More
Ammonium Molybdate Reagent I for Phosphorus by the Stannous Chloride Method (without extraction), Reagents
Ammonium Molybdate Reagent I for Phosphorus by the Stannous Chloride Method (without extraction), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$179.40
Chemical Identifiers
| CAS | 7732-18-5 , 7664-93-9 , 7664-93-9 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-883
|
Reagents Holdings Llc
CA2200001A |
1 L |
Each of 1 for $179.40
|
|
|||||
Chemical Identifiers
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| O |
Specifications
| Colorless to Light Yellow | |
| H2O | |
| O | |
| 18.02 | |
| Poly Bottle | |
| <3 | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.