Learn More
Hach Company Ammonium Molybdate Reagent, 100 mL
Supplier:  Hach Company 11032H
Encompass
Ammonium Molybdate Reagent used for Phosphate determinations by test kit methods.100 mL Bottle. For use with the Phosphorus, Orthophosphate (Reactive) Test Kit, Model PO-14 (item #147500)
Catalog No. 50-199-7378