Learn More
Strem, An Ascensus Company CAS# 12027-67-7. 500g. Ammonium molybdate tetrahydrate (ACS). MFCD00167059
SDP
Catalog No. 5090113582
Encompass
CAS# 12027-67-7. 500g. Ammonium molybdate tetrahydrate (ACS). MFCD00167059. Molecular Weight: 1163.80 (1235.86). Molecular Formula: (NH4)6Mo7O24∙4H2O. Color/form: white pwdr. Strem# 93-0245. http://www.strem.com/catalog/v/93-0245/
Catalog No. 50-901-13582 Supplier Strem, An Ascensus Company Supplier No. 930245500G
CAS# 12027-67-7. 500g. Ammonium molybdate tetrahydrate (ACS). MFCD00167059. Molecular Weight: 1163.80 (1235.86). Molecular Formula: (NH4)6Mo7O24∙4H2O. Color/form: white pwdr. Strem# 93-0245. http://www.strem.com/catalog/v/93-0245/