Learn More
ALADDIN SCIENTIFIC CORPORATION Ammonium molybdate tetrahydrat, 25g
Catalog No. 5000082531
Encompass_Preferred
Ammonium molybdate tetrahydrate is used in cryo-negative staining and helps visualize particles while preserving the sample Cryo-negative staining with Ammonium molybdate has been used to visualize Escherichia coli chaperone GroEL 20S proteasome from moss tobacco mosaic virus bacateriophage T2 tomato bushy stunt virus (TBSV) bovine liver catalase crystals turnip yellow mosaic virus and keyhole limpet hemocyanin (KLH) TYPES 1 and 2 Ammonium molybdate tetrahydrate has been used to catalytically desulfurize thiophene and its derivatives
Catalog No. 50-000-82531 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. A11637825G
Add to Cart
Add to Cart
Ammonium molybdate tetrahydrate is used in cryo-negative staining and helps visualize particles while preserving the sample Cryo-negative staining with Ammonium molybdate has been used to visualize Escherichia coli chaperone GroEL 20S proteasome from moss tobacco mosaic virus bacateriophage T2 tomato bushy stunt virus (TBSV) bovine liver catalase crystals turnip yellow mosaic virus and keyhole limpet hemocyanin (KLH) TYPES 1 and 2 Ammonium molybdate tetrahydrate has been used to catalytically desulfurize thiophene and its derivatives